Learn More
Thomson Instrument Company Ultra Yield 2.5L Flask, Sterile | CS6
Supplier:  Thomson Instrument Company 931136B
Encompass
Thomson Instrument Company Ultra Yield Flasks are designed to be either single use or autoclaved up to 3x, and feature a unique geometry that enhances gas exchange. The Ultra Yield system (flasks, Plasmid+ media and AirOtop seals) have proven over the last decade to improve yields up to 10x over traditional flasks with E. coli and other microbial cultures. These plastics flasks are designed to be fully scalable, fit standard clamp sizes and replace traditional glass flasks. All volumes,ranging from 125mL to 2.5L, can be shaken at the same speed, to optimize shaker utilization. Thomson Instrument Company manufactures its products in the USA to assure same day shipping.
Catalog No. 50996056
Only null left