Learn More
DWK Life Sciences Wheaton™ Ultra Pure OmniFlex Plus Stoppers
Featuring PTFE-like coatings to prevent reactions due to sample contact with rubber. Wheaton™ Ultra Pure OmniFlex Plus Stoppers are manufactured in a silicone-free environment.
$2,748.79 - $3,685.12
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Color | Gray |
| Material | Bromobutyl Rubber |
| Product Type | Straight Plug Stopper |
Description
- Autoclavable
- Completely coated with a fluorinated polymer
- Complies with US, European and Japanese Pharmacopeias
Specifications
| Autoclavable | |
| Gray | |
| Bromobutyl Rubber | |
| Straight Plug Stopper |