missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Ultra Physiological Fluid Kit

Catalog No. bc80211777
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BC80211777 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BC80211777 Supplier Biochrom Supplier No. 80211777
Only null left

Biochrom Amino Acid Analysis reagents and buffers are manufactured from high quality chemicals and materials and subjected to rigorous control procedures, under the ISO 13485 Quality System.

Biochrom buffers are guaranteed to give accurate and reproducible results for up to 3 years from manufacture, provided the storage conditions are adhered to. All reagents have stable chemistry and have been formulated to be stored at ambient temperature so, conveniently, do not need refrigeration.

Specifications

Product Type Fluid Kit
Content And Storage Store at ambient temperature
Quantity 1 Kit
Form Liquid