missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Ultra-High Performance Fingertight Fittings, PEEK

Catalog No. 111010761
Change view
Click to view available options
Type:
Fingertight Nut
Micro Ferrule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
111010761 Fingertight Nut
111010334 Micro Ferrule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111010761 Supplier MilliporeSigma™ Supelco™ Supplier No. 51262U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The innovative line of Upchurch Ultra High Performance fittings are desigined to withstand extreme temperature anf pressures. This line of fittings is perfect for use within the increasingly demanding requirements for today's high performance analytical systems. The fittings are a 10/32 fingertight style for use with 1/16 in. and 1/32 in. OD tubing.

Specifications

Type Fingertight Nut
Color Black
Packaging Quantity 10/Pk.
Description 10-32
For Use With 1/16 Tubing
Material PEEK