missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Ultra-High Performance Fingertight Fittings, PEEK
Specifications
Specifications
| Type | Micro Ferrule |
| Color | Black |
| Model | LiteTouch |
| Packaging Quantity | 10/Pk. |
| Description | 1/32 in. |
| For Use With | 1/32 Tubing |
| Material | PEEK |