Across International Llc ULTRA GRADE 19 VACUUM PUMP OIL
Catalog No. NC0714654
Ultra grade 19 vacuum pump oil in different packages. General use for laboratory mechanical pumps, for rapid pumping with low ultimate pressure. Good for all vacuum pumps supplied by Across International.
Catalog No.
NC0714654
Supplier
Across International LlcSupplier No.
VO1Q
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ultra grade 19 vacuum pump oil in different packages. General use for laboratory mechanical pumps, for rapid pumping with low ultimate pressure. Good for all vacuum pumps supplied by Across International.