missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Reynold's Wrap Aluminum Foil

Catalog No. 15078290
Encompass
Change view
Click to view available options
Length (English):
50 ft.
75 ft.
Width (English):
12 in.
18 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Width (English)
15078290 75 ft. 12 in.
15078291 50 ft. 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15078290 Supplier Fisherbrand™ Supplier No. 6101899

Pure aluminum foil

Sheets are ideal for covering flasks, beakers, petri dishes, weigh boats, disposable crucibles, and wrapping samples.

Specifications

Applications Ideal for covering flasks, beakers, petri dishes, weigh boats, disposable crucibles, and wrapping samples
Length (English) 75 ft.
Product Type Aluminum Foil
Width (English) 12 in.
Material Aluminum