Learn More
3M™ Ultimate FX Full Facepiece Respirator Parts and Accessories
Catalog No. 19177812
Change view
Click to view available options
Model:
FF-400-03
FF-400-04
FF-400-06
FF-400-07
FF-400-08
FF-400-09
FF-400-11
FF-400-13
FF-400-15
Product Type:
Bayonet Assembly
Exhalation Valve
Head Harness Assembly
Lens Assembly
Lens Cover
Nosecup Assembly
Speaking Diaphragm Assembly
Valve Cover
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Product Type
19177812 FF-400-07 Exhalation Valve
19177810 FF-400-03 Lens Assembly
19177811 FF-400-04 Head Harness Assembly
19177813 FF-400-08 Bayonet Assembly
19177814 FF-400-09 Valve Cover
19177815 FF-400-11 Nosecup Assembly
19177816 FF-400-13 Speaking Diaphragm Assembly
19177870 FF-400-15 Lens Cover
17623800 FF-400-06 Head Harness Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19177812 Supplier 3M™ Supplier No. 7000052738
Only null left
Replacement parts and accessories for use with the Ultimate FX full facepiece reusable respirators FF-401, 402, and 403.
  • Approved 3M replacement part
  • Designed for use with the 3M Ultimate FX Full Facepiece Reusable Respirators (sold separately)
  • Maintains original performance standards of facepiece
Specifications
Product Type Exhalation Valve
For Use With (Application) Assembly, blasting, cleaning, demolition, electrical, facility maintenance, grinding, health and safety testing, machine operation, painting, remediation, sanding, welding
For Use With (Equipment) Ultimate FX full facepiece reusable respirator FF-401, 402 and 403
Includes Mounting seat and 3M™ Cool Flow™ valve
Model FF-400-07