Learn More
New England Biolab Ltd Uracil-DNA Glycosylase (UDG) 5000 units
Supplier: New England Biolab Ltd M0280L
E. coli Uracil-DNA Glycosylase (UDG) catalyses the release of free uracil from uracil-containing DNA. UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA, but not from oligomers (6 or fewer bases).