Learn More
Enzo Life Sciences Ubiquitin monoclonal antibody (P4D1) (100 g) DFS Item
Supplier:  Enzo BMLPW09300100
Clone: P4D1. Host: Mouse. Isotype: IgG1. Immunogen: Denatured bovine ubiquitin. UniProt: P0CG53 (UBB), P0CH28 (UBC), P62992 (RPS27A), P63048 (UBA52). Source: Purified from hybridoma tissue culture supernatant. Species Reactivity: Species independent. Specificity: Recognizes mono- and poly-ubiquitin protein conjugates, free polyubiquitin chains and free ubiquitin. Applications: IHC, IP, WB. Recommended dilutions: Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified form cell culture media. Formulation: Liquid. In PBS containing 0.01% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20C.
Catalog No. 501031786
Only null left
Explore available promotions