Learn More
Thermo Scientific Chemicals Ubiquitin E1 Inhibitor, PYR-41
CAS: 418805-02-4 | C17H13N3O7·3H2O, C17H13N3O7 | 371.31 g/mol
Supplier: Thermo Scientific Chemicals J65388MB
| Quantity | 25 mg |
|---|
Description
PYR 41 is an irreversible and cell-permeable UBE1 inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity
Chemical Identifiers
| 418805-02-4 | |
| C17H13N3O7 | |
| Ubiquitin-Activating Enzyme E1 Inhibitor; PYR-41 |
Specifications
| 25 mg | |
| -30°C to -10°C | |
| 418805-02-4 | |
| 425.40g/mol | |
| Light Sensitive | |
| Signal Transduction Reagent | |
| ARGIPZKQJGFSGQ-UHFFFAOYSA-N | |
| ethyl 4-{4-[(5-nitrofuran-2-yl)methylidene]-3,5-dioxopyrazolidin-1-yl}benzoate |
| Dark Purple | |
| PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity. | |
| C17H13N3O7 | |
| Ubiquitin-Activating Enzyme E1 Inhibitor; PYR-41 | |
| Soluble in DMSO at 5mg/ml with warming | |
| Solid | |
| CCOC(=O)C1=CC=C(C=C1)N1NC(=O)C(=CC2=CC=C(O2)[N+]([O-])=O)C1=O | |
| 371.31 |
Safety and Handling
Recommended Storage : Store at -20°C