missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubenimex Hydrochloride 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 108°C |
| Color | White |
| Quantity | 25 mg |
| Formula Weight | 344.84 |
| Percent Purity | ≥97.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ubenimex Hydrochloride [for Biochemical Research] |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer (-20 °C) |