missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride, BiotechGrade, 99.9%, Spectrum™ Chemical
Description
Spectrum™ Chemical Cesium Chloride, BiotechGrade is a colorless inorganic compound and a solid. It is used in isopynic centrifugation as well as in analytical chemistry as a reagent.
Specifications
Specifications
| Color | Fine, white, crystal |
| CAS Max % | 100% |
| Quantity | 100 g |
| Packaging | Poly Bottle |
| Percent Purity | 99.9% |
| Grade | Biotech |