Learn More
Selleck Chemical Llc U0126-EtOH 25mg 1173097-76-1
Supplier: Selleck Chemicals Co Ltd S110225MG
U0126-EtOH is a highly selective inhibitor of MEK1/2 with IC50 of 0.07 ?M/0.06 ?M in cell-free assays, 100-fold higher affinity for ?N3-S218E/S222D MEK than PD98059. U0126 inhibits autophagy and mitophagy with antiviral activity. *For Research & Development use only. Product is not intended for drug, household, or human consumption.