Learn More
Ibidi Gmbh -Slide I, ibiTreat - Tissue Culture Treated Polymer Coverslip
Catalog No. 50305739
Encompass
The ibidi product family is comprised of a variety of -Slides and -Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The -Slide I combines the features of a cell culture dish with those of a glass cover slip. One flow through observation channel is integrated in the -Slide. Its large observation area and high-end optical quality permits convenient monitoring of a vast variety of cellular assays. #1.5 polymer coverslip, tissue culture treated, sterile, 15 pcs./box
Catalog No. 50305739 Supplier Ibidi Gmbh Supplier No. 80106
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The ibidi product family is comprised of a variety of -Slides and -Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The -Slide I combines the features of a cell culture dish with those of a glass cover slip. One flow through observation channel is integrated in the -Slide. Its large observation area and high-end optical quality permits convenient monitoring of a vast variety of cellular assays. #1.5 polymer coverslip, tissue culture treated, sterile, 15 pcs./box