The ibidi product family is comprised of a variety of -Slides and -Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The Slide 2 Well CoCulture is comprised of two arrays of 33 square fields whereby cells can be cultivated and investigated with microscopical methods. It is intended for investigating experimental parameters such as antibody dilution, seeding density or most effective drug concentrations. A special application is the co-cultivation of different cell types which share one medium supernatant but grow separately from each other. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box