missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ U-Shaped Magnet

Catalog No. S95958
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95958 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95958 Supplier Eisco™ Supplier No. PH0795D

Includes keepers.

  • Alnico horshoe magnet
  • Cardboard box

Order Info

Dimensions: 3 in. (7.6cm), 0.39 x 0.43 in. (10 x 9mm) section

Specifications

Shape U-shaped
For Use With (Application) Used to view Magnetic fields
Grade 9 to 12
Product Type Horseshoe Magnet
Includes Keepers