Learn More
MCR Safety Double Matrix Headgear
Description
- Double Matrix Head Gear
- Ratchets to fit
- Includes headgear only. Plastic Face Shield must be purchased separately (494400, 494700, 494800).
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1+, CSA Z94.1 |
| Description | Ratchet |
| Lens Tint | Clear |
| For Use With (Equipment) | Double matrix headgear |
| Includes | Headgear only. Plastic Face Shield must be purchased separately (494400, 494700, 494800). |
| No. per Case | 60/Cs. |
| Lens Material | Polycarbonate |