missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ U-Form Glass Absorption Tube
Made from High Quality, Borosilicate 3.3 Glass
Supplier: Eisco™ CH0040B
Description
- Strong walls made of borosilicate 3.3 glass
- Autoclavable, with superior chemical durability
- Low temperature gradient can withstand high temperatures & thermal shocks
Specifications
| 35 mL | |
| Clear | |
| 125 mm |
| Borosilicate Glass (3.3) | |
| Autoclavable | |
| 15 mm |