missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals U-18666A, 95%

Catalog No. AAJ67389MA
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67389MA 10 mg
AAJ67389LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67389MA Supplier Thermo Scientific Chemicals Supplier No. J67389MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3039-71-2 | C25H41NO2.HCl, C25H42ClNO2 | 424.07 g/mol

Impaired trafficking of cholesterol to or from the endoplasmic reticulum (ER) has been implicated in many disease states including Neimann-Pick disease type C (NPC), Alzheimers disease and atherosclerosis. U-18666A is a cell permeable drug that inhibits cholesterol trafficking. It inhibits cholesterol transport from late endosomes/lysosomes to the endoplasmic reticulum (ER), but not cholesterol transport to the plasma membrane as demonstrated in many cell types including macrophages, primary cortical neurons and primary fibroblasts. In macrophages, micromolar concentrations of U-18666A inhibit multiple pathways of cholesterol trafficking from late endosomes, whereas nanomolar concentrations impair cholesterol trafficking to the ER, a response similar to that found in NPC. U-18666A inhibits oxidosqualene cyclase at high (>0.5 mM) concentrations and oral doses (10 mg/kg) induces cataracts in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3039-71-2
Molecular Formula C25H41NO2.HCl
Synonym 3β-[2-(Diethylamino)ethoxy]androstenone hydrochloride

Specifications

Quantity 10 mg
Description White to off-white
Content And Storage 2°C to 8°C
Formula Weight 424.06
Melting Point 195°C to 196.5°C
Solubility Soluble in water at 10mg/ml
Product Type U-18666A, 95%
Form Solid
Assay 95%

RUO – Research Use Only