TA Instruments Waters LLC TZERO LIDS (pkg. of 100) Temperature Range: -180C to 600C
Catalog No. NC9553079
Lightweight aluminum lids for use in sample encapsulation with the Tzero Pans (P/N: 901683.901) and the Tzero Low-Mass Pans (P/N: 901670.901). The seal is not hermetic. 100/pack
Catalog No.
NC9553079
Supplier
TA Instruments Waters LLCSupplier No.
901671.901
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Lightweight aluminum lids for use in sample encapsulation with the Tzero Pans (P/N: 901683.901) and the Tzero Low-Mass Pans (P/N: 901670.901). The seal is not hermetic. 100/pack