missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 258 Coveralls
GREENER_CHOICE

Catalog No. p-200007049
Change view
Click to view available options
Size:
6X-Large
7X-Large
Height (English):
5.8 to 6 ft.
6.1 to 6.5 ft.
Height (Metric):
176.78 to 182.88 cm
185.92 to 198.12 cm
Catalog No. Size Height (English) Height (Metric)
17988052 6X-Large 5.8 to 6 ft. 176.78 to 182.88 cm
17988054 7X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
2 missing translation for 'options'

Catalog No. 17988052

Supplier: DuPont™ IC253BWH6X0025CS

Only null left

Coveralls create an effective barrier to dry particles, microorganisms, and nonhazardous liquids in cleanroom environments.

  • Garments are gamma sterilized to SAL of 10-6
  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort
  • Full traceability on all sterilized apparel with certificates of sterility available

Specifications

Product Type Cleanroom Coveralls
Size 6X-Large
Material Tyvek
Color White
Ankle Style Elastic
Cuff Style Elastic
Closure Type Zipper
Seam Type Serged
Sterility Sterile
Collar Type Bound
Description Elastic wrist and ankle, raglan sleeves, bound neck
Model IC258SWH
Size (English) Chest 38 to 40 in.
Size (English) Inseam 34 in.
Size (Metric) Chest 96.52 to 101.6 cm
Size (Metric) Inseam 86.36 cm
Sleeve Style Raglan
Thickness 5.4 mil
For Use With (Application) Barrier to dry particles, microorganisms and non-hazardous liquids
Length (English) Sleeve 38 in.
Length (Metric) Sleeve 96.52 cm
Height (English) 5.8 to 6 ft.
Height (Metric) 176.78 to 182.88 cm
Show More Show Less