missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
GREENER_CHOICE

Catalog No. 19188130
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Height (English):
5 to 5.7 ft.
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
6.3 to 7 ft.
6.4 to 7.2 ft.
Height (Metric):
152.4 to 173.7 cm
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
192.02 to 213.3 cm
195.07 to 219.45 cm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
19188130 5X-Large 6.4 to 7.2 ft. 195.07 to 219.45 cm
17990511 Small 5 to 5.7 ft. 152.4 to 173.7 cm
17988034 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
17988032 Large 5.8 to 6 ft. 176.78 to 182.88 cm
17988035 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
17988031 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
17988036 4X-Large 6.3 to 7 ft. 192.02 to 213.3 cm
17988033 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19188130 Supplier DuPont™ Supplier No. IC253BWH5X00250C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Clean-processed coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.

  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam
  • Elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort

Specifications

Product Type Cleanroom Coveralls
Size 5X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Particle, Non-toxic Particulates
Collar Type Bound
Size (English) Chest 58 to 62 in.
Size (English) Inseam 38 in.
Size (Metric) Chest 147 to 157 cm
Size (Metric) Inseam 96.5 cm
Sleeve Style Dolman
Thickness 5 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
Length (English) Sleeve 42.5 in.
Length (Metric) Sleeve 11 cm
Height (English) 6.4 to 7.2 ft.
Height (Metric) 195.07 to 219.45 cm
Packaging Clean-processed
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less