Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)
Clean-processed coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.
Supplier:  DuPont™ IC253BWH3X00250C
 View more versions of this product
Catalog No. 17-988-035
Only null left
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam
  • Elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Covered Elastic
Collared
Bound
Bound
35 in.
88.9 cm
5 mil
40.75 in.
6.2 to 6.6 ft.
Clean-processed
3X-Large
Tyvek
Single-use
Covered Elastic
Zipper
Non-sterile
46 to 52 in.
116.8 to 132.08 cm
Dolman
Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
103.5 cm
188.97 to 201.16 cm
Class 1000 (ISO 6)
Product Suggestions
Product Suggestions