Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)
Clean-processed coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.
Supplier:  DuPont™ IC253BWHLG00250C
 View more versions of this product
Catalog No. 17-988-032
Only null left
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam
  • Elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Covered Elastic
Collared
Bound
Bound
24.25 in.
61.5 cm
5 mil
36.25 in.
5.8 to 6 ft.
Clean-processed
Large
Tyvek
Single-use
Covered Elastic
Zipper
Non-sterile
38 to 40 in.
96.52 to 101.6 cm
Dolman
Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
92.7 cm
176.78 to 182.88 cm
Class 1000 (ISO 6)
Product Suggestions
Product Suggestions