missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
GREENER_CHOICE

Catalog No. 17988031
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
Small
X-Large
Height (English):
5 to 5.7 ft.
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
6.3 to 7 ft.
6.4 to 7.2 ft.
Height (Metric):
152.4 to 173.7 cm
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
192.02 to 213.3 cm
195.07 to 219.45 cm
Catalog No. Size Height (English) Height (Metric)
17988031 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
17990511 Small 5 to 5.7 ft. 152.4 to 173.7 cm
17988034 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
17988032 Large 5.8 to 6 ft. 176.78 to 182.88 cm
17988035 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
19188130 5X-Large 6.4 to 7.2 ft. 195.07 to 219.45 cm
17988036 4X-Large 6.3 to 7 ft. 192.02 to 213.3 cm
17988033 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
8 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Catalog No. 17988031

Supplier: DuPont™ IC253BWHMD00250C

Only null left
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Clean-processed coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.

  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam
  • Elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort

Specifications

Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Collar Type Bound
Size (English) Chest 36 to 38 in.
Size (English) Inseam 23.5 in.
Size (Metric) Chest 91.44 to 96.52 cm
Size (Metric) Inseam 59.6 cm
Sleeve Style Dolman
Thickness 5 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
Length (English) Sleeve 35.5 in.
Length (Metric) Sleeve 90.17 cm
Height (English) 5.5 to 5.10 ft.
Height (Metric) 167.64 to 155.44 cm
Packaging Clean-processed
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less