Learn More
DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls
Tyvek IsoClean delivers an ideal balance of protection, durability, and comfort. Bulk packaged in double transparent poly liners.
Supplier:  DuPont™ IC182BWHSM002500
 View more versions of this product
Catalog No. 190881120A
Only null left
Explore available promotions
Description
Description
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing.
  • Raglan sleeve design for greater range of motion and comfort.
  • Bulk packaged in double transparent poly liners.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex
  • D13471798,D13471822,D13471855,D13471932,D13471962,D13471992
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Elastic
Collared
Bound
Bound
31.5 in.
80 cm
5 mil
36 in.
5 to 5.7 ft.
Bulk
Small
Tyvek
Single-use
Elastic
Zipper
Non-sterile
32 to 36 in.
81.28 to 91.44 cm
Raglan
Premium Protection For Controlled Environments And Inherent Barrier To Dry Particles, Microorganisms, And Non-hazardous Liquids
91.44 cm
152.4 to 173.73 cm
Class 1000 (ISO 6)
Product Suggestions
Product Suggestions