Learn More
DuPont™ Tyvek™ IsoClean™ 265 Series Lab Coat
GREENER_CHOICE
Catalog No. 17001075
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001075 Small
17001077 Large
17001078 X-Large
17001079 2X-Large
17001080 3X-Large
17001081 4X-Large
17001076 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17001075 Supplier DuPont™ Supplier No. IC265SWHSM00300B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tyvek™ IsoClean™ lab coats provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in controlled environments.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Knit cuff for tighter fit at wrist.
  • Front zipper closure for easy donning and doffing.
  • Raglan sleeve design for greater range of motion and comfort.
  • Three front pockets.
  • A-line silhouette.
  • Bulk packaged in double transparent poly liners.
  • Manufactured under specifications that do not contain natural rubber latex
  • D15541914,D15541920,D15541915,D15541916,D15541917,D15541918,D15541919
Specifications
Product Type Controlled Environments Lab Coat
Size Small
Gender Unisex
Material Tyvek
Color White
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
No. of Pockets 3
Seam Type Serged
Sleeve Style Raglan
Packaging Type Bulk Packaged, Double Transparent Poly Liners
Show More Show Less