missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Pig Tyvek™ IsoClean™ Cleanroom Lab Coat
Description
- The lab coat is made from Tyvek™ material, which is a flashspun high-density polyethylene.
- This provides an excellent barrier against dry particulates, microorganisms, and non-hazardous liquid splashes.
- The unique manufacturing process for the Tyvek™ IsoClean™ fabric is designed to minimize both linting and particle shedding, which is a critical requirement for maintaining the integrity of ISO Class 5-8 cleanroom environments.
- The Tyvek™ material is known for its high durability and resistance to tearing and abrasion, ensuring the lab coat remains intact and protective during use.
- The nonwoven structure of Tyvek™ allows air and moisture vapor to pass through, which helps to reduce heat stress and keeps the wearer more comfortable during extended periods of use.
- These lab coats are available in both clean-processed and sterile options. The sterile versions are gamma-irradiated to a Sterility Assurance Level (SAL) of -6, making them suitable for sterile compounding and other controlled applications.
- The lab coat typically features a front zipper or snap closure, a collar, and elastic wrists to provide a secure fit and a professional appearance while preventing contamination.
Spécifications
Spécifications
| Product Type | Lab Coat |
| Size | Large |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Closure Type | Zipper |
| Cuff Style | Knit |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Microorganisms, Dry Particulates, Splash |
| Afficher plus de résultats |