Learn More
New Pig Tyvek™ IsoClean™ Cleanroom Lab Coat
Catalog No. 17987878
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17987878 3X-Large
17987875 Large
17987874 Medium
17987873 Small
17987876 X-Large
17987877 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17987878 Supplier New Pig Supplier No. WPL1016XXXL
Only null left
Lab coat provides an excellent barrier to particles, microorganisms and non-hazardous light liquid splash in ISO Class 5-8 cleanrooms
  • The lab coat is made from Tyvek™ material, which is a flashspun high-density polyethylene.
  • This provides an excellent barrier against dry particulates, microorganisms, and non-hazardous liquid splashes.
  • The unique manufacturing process for the Tyvek™ IsoClean™ fabric is designed to minimize both linting and particle shedding, which is a critical requirement for maintaining the integrity of ISO Class 5-8 cleanroom environments.
  • The Tyvek™ material is known for its high durability and resistance to tearing and abrasion, ensuring the lab coat remains intact and protective during use.
  • The nonwoven structure of Tyvek™ allows air and moisture vapor to pass through, which helps to reduce heat stress and keeps the wearer more comfortable during extended periods of use.
  • These lab coats are available in both clean-processed and sterile options. The sterile versions are gamma-irradiated to a Sterility Assurance Level (SAL) of -6, making them suitable for sterile compounding and other controlled applications.
  • The lab coat typically features a front zipper or snap closure, a collar, and elastic wrists to provide a secure fit and a professional appearance while preventing contamination.
Specifications
Product Type Lab Coat
Size 3X-Large
Gender Unisex
Material Tyvek
Color White
Closure Type Zipper
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Microorganisms, Dry Particulates, Splash
No. of Pockets 3
Latex Free Yes
Autoclavable Not Autoclavable
Cleanroom Class Class 100-100,000 (ISO 5-8)
Show More Show Less