missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Pig Tyvek™ IsoClean™ Cleanroom Frock
Frock provides an excellent barrier to particles, microorganisms and non-hazardous light liquid splash in ISO Class 5-8 cleanrooms
Supplier: New Pig WPL1015XXXL
Description
- Constructed from Tyvek™ material (flashspun high-density polyethylene), this frock provides an inherent barrier to particles, microorganisms, and non-hazardous liquid splashes, making it ideal for controlled environments.
- The fabric is specially processed to minimize both linting and particle shedding, which is a critical feature for maintaining the integrity of ISO Class 5-8 cleanrooms.
- Tyvek™ is known for its strength and resistance to tearing and abrasion, allowing the frock to withstand the rigors of repeated use without compromising its protective qualities.
- The frocks are available in both clean-processed and sterile versions.
- The sterile options are gamma-irradiated to a Sterility Assurance Level (SAL) of -6, making them suitable for sterile environments like those in pharmaceutical manufacturing.
- The unique nonwoven structure of Tyvek™ allows air and moisture vapor to pass through, providing a high degree of comfort and reducing heat stress for the wearer.
- The frocks are designed for ease of donning and doffing with features like a front zipper with a placket for increased protection, and an extended length to ensure full coverage over the user's clothing.
Specifications
| Cleanroom Frock | |
| Tyvek | |
| Single-use | |
| Zipper | |
| Serged | |
| Yes | |
| Raglan Sleeve |
| 3X-Large | |
| White | |
| Non-sterile | |
| Elastic | |
| Protection against Particles, Microorganisms | |
| None | |
| Class 100-100,000 (ISO 5-8) |