missing translation for 'onlineSavingsMsg'
Learn More

New Pig Tyvek™ IsoClean™ Cleanroom Frock

Catalog No. 17987869
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17987869 Large
17987868 Medium
17987867 Small
17987870 X-Large
17987871 2X-Large
17987872 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17987869 Supplier New Pig Supplier No. WPL1015L
Only null left

Frock provides an excellent barrier to particles, microorganisms and non-hazardous light liquid splash in ISO Class 5-8 cleanrooms

  • Constructed from Tyvek™ material (flashspun high-density polyethylene), this frock provides an inherent barrier to particles, microorganisms, and non-hazardous liquid splashes, making it ideal for controlled environments.
  • The fabric is specially processed to minimize both linting and particle shedding, which is a critical feature for maintaining the integrity of ISO Class 5-8 cleanrooms.
  • Tyvek™ is known for its strength and resistance to tearing and abrasion, allowing the frock to withstand the rigors of repeated use without compromising its protective qualities.
  • The frocks are available in both clean-processed and sterile versions.
  • The sterile options are gamma-irradiated to a Sterility Assurance Level (SAL) of -6, making them suitable for sterile environments like those in pharmaceutical manufacturing.
  • The unique nonwoven structure of Tyvek™ allows air and moisture vapor to pass through, providing a high degree of comfort and reducing heat stress for the wearer.
  • The frocks are designed for ease of donning and doffing with features like a front zipper with a placket for increased protection, and an extended length to ensure full coverage over the user's clothing.

Specifications

Product Type Cleanroom Frock
Size Large
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Seam Type Serged
For Use With (Application) Protection against Particles, Microorganisms
Latex Free Yes
No. of Pockets None
Sleeve Style Raglan Sleeve
Cleanroom Class Class 100-100,000 (ISO 5-8)
Show More Show Less