Learn More
New Pig Tyvek™ IsoClean™ Cleanroom Boot Cover
Catalog No. p-200136142
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Packaging Quantity:
100 Pk.
300 Pk.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Quantity
17987861 Large 100 Pk.
17987860 Medium 100 Pk.
17987859 Small 100 Pk.
17987862 X-Large 100 Pk.
17987865 Large 300 Pk.
17987864 Medium 300 Pk.
17987863 Small 300 Pk.
17987866 X-Large 300 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17987861 Supplier New Pig Supplier No. WPL1012L
Only null left
These boot covers provide an excellent barrier to particles, microorganisms and non-hazardous light liquid splash in ISO Class 5-8 cleanrooms
  • Made from Tyvek™ material, which is a patented flashspun high-density polyethylene, these boot covers provide an inherent barrier to particles, microorganisms, and non-hazardous liquid splashes.
  • The fabric is specially processed to be exceptionally low-linting and to minimize particle shedding, which is a critical feature for maintaining the integrity of ISO Class 5-8 cleanrooms.
  • Tyvek™ is known for its durability. The material has a high abrasion resistance and is strong enough to withstand rigorous use without wearing away, ensuring reliable protection.
  • The boot covers are available in both clean-processed and sterile options.
  • The sterile versions are gamma-irradiated to a Sterility Assurance Level (SAL) of 10-6, making them suitable for GMP Class A/B (ISO Class 5) environments.
  • They are designed with features like tunnelled elastication at the shin and ankle, along with ankle ties, to provide a secure fit over footwear and prevent the boot cover from slipping or riding up.
  • The boot covers are often folded to facilitate aseptic donning, minimizing the risk of contamination during the gowning process.
Specifications
Size Large
Material Tyvek
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top with Ankle Ties
Sole Type PVC
Thread Color White
Packaging Quantity 100 Pk.
Resistance Microorganisms, Dry Particulates, Splash
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100 (ISO 5)
Show More Show Less