missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Pig Tyvek™ IsoClean™ Cleanroom Boot Cover
Description
- Made from Tyvek™ material, which is a patented flashspun high-density polyethylene, these boot covers provide an inherent barrier to particles, microorganisms, and non-hazardous liquid splashes.
- The fabric is specially processed to be exceptionally low-linting and to minimize particle shedding, which is a critical feature for maintaining the integrity of ISO Class 5-8 cleanrooms.
- Tyvek™ is known for its durability. The material has a high abrasion resistance and is strong enough to withstand rigorous use without wearing away, ensuring reliable protection.
- The boot covers are available in both clean-processed and sterile options.
- The sterile versions are gamma-irradiated to a Sterility Assurance Level (SAL) of 10-6, making them suitable for GMP Class A/B (ISO Class 5) environments.
- They are designed with features like tunnelled elastication at the shin and ankle, along with ankle ties, to provide a secure fit over footwear and prevent the boot cover from slipping or riding up.
- The boot covers are often folded to facilitate aseptic donning, minimizing the risk of contamination during the gowning process.
Specifications
Specifications
| Size | Large |
| Material | Tyvek |
| Latex Free | Yes |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Top with Ankle Ties |
| Sole Type | PVC |
| Thread Color | White |
| Packaging Quantity | 100 Pk. |
| Show More |