missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Frock
GREENER_CHOICE

Catalog No. 17990524
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17990524 2X-Large
17990522 Large
17990523 X-Large
17990526 4X-Large
17990525 3X-Large
17990521 Medium
17990520 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 17990524 Supplier DuPont™ Supplier No. IC264SWH2X00300C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Should not be worn around open flame, heat source, sparks or ignition sources.

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Clean processed garments offer lowest level of particle shedding within DuPont product portfolio.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette

Specifications

Size 2X-Large
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Collar Type Bound
Size (English) Chest 40 in to 43 in.
Size (Metric) Chest 101.6 to 109.2 cm
Sleeve Style Raglan Sleeve
Packaging Type Individually Packaged
Length (English) 53.75 in.
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less