Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Bag
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. These filling needle bags stay in place via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically removed with forceps after the filling needles are set in place, without having to cut, or remove particulate generating autoclave tape.
$3695.15 - $8500.93
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Drawstring
Color White
Disposable Single-use
View More Specs
Products 27
Catalog Number Mfr. No. Dimensions (L x W) Capacity (English) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Capacity (English) Price Quantity  
19-990-291
View Documents
Keystone Cleanroom Products
BGT2A0103D10
3 x 1 in. 3 oz.
Case of 500 Each for $4,456.24
Only null left
19-990-292
View Documents
Keystone Cleanroom Products
BGT2A0104D
4 x 1 in. 4 oz.
Case of 500 Each for $3,874.38
Only null left
19-990-293
View Documents
Keystone Cleanroom Products
BGT2A0105D
5 x 1 in. 5 oz.
Case of 500 Each for $3,945.34
Only null left
19-990-294
View Documents
Keystone Cleanroom Products
BGT2A0106D
6 x 1 in. 6 oz.
Case of 500 Each for $3,945.34
Only null left
19-990-295
View Documents
Keystone Cleanroom Products
BGT2A0107D
7 x 1 in. 7 oz.
Case of 500 Each for $4,030.49
Only null left
19-990-296
View Documents
Keystone Cleanroom Products
BGT2A0108D
8 x 1 in. 8 oz.
Case of 500 Each for $4,101.44
Only null left
19-990-297
View Documents
Keystone Cleanroom Products
BGT2A0111D
11 x 1 in. 11 oz.
Case of 500 Each for $4,328.51
Only null left
19-990-310
View Documents
Keystone Cleanroom Products
BGT2A02503D
3 x 2.5 in. 7.5 oz.
Case of 500 Each for $5,321.94
Only null left
19-990-298
View Documents
Keystone Cleanroom Products
BGT2A0202D16
2 x 2 in. 4 oz.
Case of 500 Each for $4,484.63
Only null left
19-990-299
View Documents
Keystone Cleanroom Products
BGT2A0203D
3 x 2 in. 6 oz.
Case of 500 Each for $4,711.69
Only null left
19-990-300
View Documents
Keystone Cleanroom Products
BGT2A0204D
4 x 2 in. 8 oz.
Case of 500 Each for $5,321.94
Only null left
19-990-301
View Documents
Keystone Cleanroom Products
BGT2A0205D
5 x 2 in. 10 oz.
Case of 500 Each for $5,392.90
Only null left
19-990-302
View Documents
Keystone Cleanroom Products
BGT2A0206D
6 x 2 in. 10 oz.
Case of 500 Each for $5,549.02
Only null left
19-990-303
View Documents
Keystone Cleanroom Products
BGT2A0207D
7 x 2 in. 14 oz.
Case of 500 Each for $5,080.68
Only null left
19-990-304
View Documents
Keystone Cleanroom Products
BGT2A0208D
8 x 2 in. 16 oz. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-990-305
View Documents
Keystone Cleanroom Products
BGT2A0209D
9 x 2 in. 18 oz. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-990-306
View Documents
Keystone Cleanroom Products
BGT2A0210D
10 x 2 in. 20 oz. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-990-307
View Documents
Keystone Cleanroom Products
BGT2A0211D
11 x 2 in. 22 oz. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-990-308
View Documents
Keystone Cleanroom Products
BGT2A0212D
12 x 2 in. 24 oz.
Case of 250 Each for $4,101.44
Only null left
19-990-309
View Documents
Keystone Cleanroom Products
BGT2A0213D
13 x 2 in. 26 oz. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-990-311
View Documents
Keystone Cleanroom Products
BGT2A0303D18
3 x 3 in. 9 oz.
Case of 500 Each for $5,549.02
Only null left
19-990-312
View Documents
Keystone Cleanroom Products
BGT2A0305D
5 x 3 in. 15 oz.
Case of 500 Each for $5,080.68
Only null left
19-990-313
View Documents
Keystone Cleanroom Products
BGT2A0306D
6 x 3 in. 18 oz.
Case of 500 Each for $5,392.90
Only null left
19-990-314
View Documents
Keystone Cleanroom Products
BGT2A0308D
8 x 3 in. 24 oz.
Case of 500 Each for $6,301.18
Only null left
19-990-315
View Documents
Keystone Cleanroom Products
BGT2A0309D
9 x 3 in. 27 oz.
Case of 500 Each for $6,372.15
Only null left
19-990-316
View Documents
Keystone Cleanroom Products
BGT2A0310D
10 x 3 in. 30 oz.
Case of 250 Each for $3,695.15
Only null left
19-990-317
View Documents
Keystone Cleanroom Products
BGT2A0312D
12 x 3 in. 36 oz.
Case of 500 Each for $6,599.21
Only null left
Description
Description
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Autoclavable
Drawstring
Single-use
Pharmaceutical Equipment
Non-sterile
Steam
<125°C
COC
White
Steam Sterilization
Tyvek
Yes
<250°F
Autoclave Bag
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'