missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 800 Hooded Coveralls

Catalog No. 19715365
Change view
Click to view available options
Size:
4X-Large
5X-Large
6X-Large
7X-Large
Small
Medium
Large
X-Large
2X-Large
3X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19715365 Small
19715361 Medium
19715360 Large
19715359 X-Large
19715356 2X-Large
19715347 3X-Large
19088821 7X-Large
19088820 6X-Large
19088819 5X-Large
19088818 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Does not provide protection from ionizing radiation
Catalog No. 19715365 Supplier DuPont™ Supplier No. TJ198TWHSM0025PI
Does not provide protection from ionizing radiation

Providing a suitable protective barrier against many low-concentration, water-based, inorganic chemicals and particles, down to one micron in size, Tyvek 800 coveralls offer an optimum fit with full freedom of movement.

Serged and over-taped seams that provide the performance of Type 3/4/5/6 garments, which have been tested to standards against pressurized liquids, heavy liquid aerosols and airborne solid particles. The coveralls provide an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size), as well as oil repellency.

  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603) and resistance to penetration by blood-borne pathogens using Phi-X174 bacteriophage (ISO 16604)
  • EN 1073-2 (protection against radioactive contamination)
  • Fabric and seams offer chemical permeation barrier to low concentration water-based inorganic chemicals
  • Serged and over-taped seams for protection and strength
  • Self-adhesive chin flap for tight seal of suit to the mask
  • Elastic face, wrists and ankles as well as glued-in waist elastic
  • Elastic thumb loops keep sleeves in place
  • Distinct orange taping of the garment seams providing
  • color contrast

Compliance

CE certified.

Specifications

Product Type Hooded Coveralls
Size Small
Color White
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Protection Class
  • Protection against biological hazards (according to EN 14126)
  • Protection against radioactive contamination (EN 1073-2) up to Class 2**
  • Protection against electrostatic discharge (antistatic according to EN 1149-5)
** Does not provide protection from ionizing radiation.
Certifications/Compliance CE certified
Size (Metric) Chest 84 to 92 cm
For Use With (Application) Industrial cleaning, Petrochemical installations, sewers. Maintenance
Height (Metric) 162 to 170 cm
Show More Show Less