missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 800 Hooded Coveralls

Catalog No. 19715356
Change view
Click to view available options
Size:
4X-Large
5X-Large
6X-Large
7X-Large
Small
Medium
Large
X-Large
2X-Large
3X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19715356 2X-Large
19715365 Small
19715361 Medium
19715360 Large
19715359 X-Large
19715347 3X-Large
19088821 7X-Large
19088820 6X-Large
19088819 5X-Large
19088818 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Does not provide protection from ionizing radiation
Catalog No. 19715356 Supplier DuPont™ Supplier No. TJ198TWH2X0025PI
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Does not provide protection from ionizing radiation

Providing a suitable protective barrier against many low-concentration, water-based, inorganic chemicals and particles, down to one micron in size, Tyvek 800 coveralls offer an optimum fit with full freedom of movement.

  • Serged and over-taped seams provide the performance of Type 3/4/5/6 garments, tested to standards against pressurized liquids, heavy liquid aerosols and airborne solid particles. The coveralls provide an effective barrier against many water-based inorganic chemicals in low concentration and particles down to 1.0 micron in size, as well as oil repellency.
  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • Protection against infective agents (EN 14126), including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603) and resistance to penetration by blood-borne pathogens using Phi-X174 bacteriophage (ISO 16604)
  • EN 1073-2 protection against radioactive contamination
  • Fabric and seams offer a chemical permeation barrier to low concentration water-based inorganic chemicals
  • Serged and over-taped seams for protection and strength
  • Self-adhesive chin flap for a tight seal of suit to the mask
  • Elastic face, wrists and ankles, as well as glued-in waist elastic
  • Elastic thumb loops keep sleeves in place
  • Distinct orange taping of the garment seams provides color contrast

Compliance

CE certified.

Specifications

Product Type Hooded Coveralls
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Protection Class
  • Protection against biological hazards (according to EN 14126)
  • Protection against radioactive contamination (EN 1073-2) up to Class 2**
  • Protection against electrostatic discharge (antistatic according to EN 1149-5)
** Does not provide protection from ionizing radiation.
Resistance Liquid, Particle
Certifications/Compliance CE certified
Size (Metric) Chest 116 to 124 cm
For Use With (Application) Industrial cleaning, Petrochemical installations, sewers. Maintenance
Height (Metric) 186 to 194 cm
Show More Show Less