missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 600 Hooded Coverall
DuPont™ Tyvek™ 600 Coverall with Hood provides an effective barrier against low concentration water-based inorganic chemicals, small sized hazardous particles down to one micron, and biological hazards.
Supplier: DuPont™ TY198TWH5X0025PI
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Elastic thumb loops keep sleeves in place
- Packaged Individually
- Products made after January, 2023 are manufactured under specifications that do not contain natural rubber latex.
- Chemical protective clothing, Category III, Type 4-B, 5-B and 6-B
- EN 1073-2 (protection against radioactive contamination)
- Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603)
- Antistatic treatment (EN 1149-5) - on both sides.
- Fabric and seams offer chemical permeation barrier to low concentration water-based inorganic chemicals
- Serged and over-taped seams for protection and strength
- Self-adhesive chin flap for tight seal of suit to the mask
- Elastic face, wrists and ankles as well as glued-in waist elastic
Certifications
Chemical Protective Clothing, Cat.III,Type 5-B,6-B
Specifications
| 5X-Large | |
| Tyvek | |
| Single-use | |
| Elastic | |
| Zipper | |
| Non-sterile | |
| Chemical Protective Clothing, Cat.III,Type 5-B,6-B | |
| Yes | |
| 25/Cs. |
| Unisex | |
| White | |
| Elastic | |
| Hooded | |
| Serged, Taped | |
| Liquid, Particle | |
| Packaged Individually | |
| None |