Learn More
DuPont™ Tyvek™ 500 Hooded Coveralls
Description
- Chemical protective clothing, Category III, Type 5-B and 6-B.
- EN 1073-2 (protection against radioactive contamination)
- Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603)
- Antistatic treatment (EN 1149-5) on both sides
- Serged external seams featuring patent-pending technology for enhanced liquid spray tightness
- Very low inward leakage due to optimized design
- Tyvek™ auto-lock zipper and storm flap for increased protection
- Elastic face, wrists and ankles as well as glued-in waist elastic
Specifications
Specifications
| Size | X-Large |
| Material | Tyvek |
| Color | White |
| Closure Type | Storm Flap |
| Model | TY198SWH |
| Quantity | 25 Coveralls/Cs. |