Learn More
DuPont™ Tyvek™ 500 Hooded Coveralls
Catalog No. 19067050
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19067050 Medium
19067049 Large
19067051 X-Large
19067047 2X-Large
19067048 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19067050 Supplier DuPont™ Supplier No. TY198SWHMD0025LA
Only null left
Tyvek™ 500 Hooded Anti-static Coveralls boast stormflaps and serged external seams, with additional protection against blood and bodily fluids.
  • Chemical protective clothing, Category III, Type 5-B and 6-B.
  • EN 1073-2 (protection against radioactive contamination)
  • Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603)
  • Antistatic treatment (EN 1149-5) on both sides
  • Serged external seams featuring patent-pending technology for enhanced liquid spray tightness
  • Very low inward leakage due to optimized design
  • Tyvek™ auto-lock zipper and storm flap for increased protection
  • Elastic face, wrists and ankles as well as glued-in waist elastic
Specifications
Size Medium
Material Tyvek
Color White
Waist Style Elastic (Glued-in)
Ankle Style Elastic
Cuff Style Elastic
Hood Type Elastic
Closure Type Storm Flap
Seam Type Serged
Model TY198SWH
Quantity 25 Coveralls/Cs.
Show More Show Less