Learn More
DuPont™ Tyvek™ 500 Hooded Coveralls
Tyvek™ 500 Hooded Anti-static Coveralls boast stormflaps and serged external seams, with additional protection against blood and bodily fluids.
Supplier: DuPont™ TY198SWHLG0025LA
Description
- Chemical protective clothing, Category III, Type 5-B and 6-B.
- EN 1073-2 (protection against radioactive contamination)
- Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603)
- Antistatic treatment (EN 1149-5) on both sides
- Serged external seams featuring patent-pending technology for enhanced liquid spray tightness
- Very low inward leakage due to optimized design
- Tyvek™ auto-lock zipper and storm flap for increased protection
- Elastic face, wrists and ankles as well as glued-in waist elastic
Specifications
| Large | |
| White | |
| Elastic | |
| Elastic | |
| Serged | |
| 25 Coveralls/Cs. |
| Tyvek | |
| Elastic (Glued-in) | |
| Elastic | |
| Storm Flap | |
| TY198SWH |