Learn More
DuPont™ Tyvek™ 400 Shirt
400 Shirt extends to the hip and offers convenient snap closures.
Supplier: DuPont™ TY303SWHLG0050VP
Description
- Shirt design with front snap closures for easy donning and doffing
- Laydown collar
- Open wrists
- Typically worn with TY350S WH pants
- Manufactured under specifications that do not contain natural rubber latex
- D15221840,D15221856,D15221820,D15221819,D15221838
Specifications
| Large | |
| Tyvek | |
| Serged | |
| Open |
| Unisex | |
| White | |
| Front Snaps | |
| 50 Pk. |