missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 400 SFR White Coverall ST120, Standard Fit Hood
Tyvek™ 400 SFR Coverall. Relaxed Fit Design. Standard Fit Hood. Storm flap. Red Zipper. Elastic Wrists and Ankles. Red External Serged Seams. White.
$291.85 - $446.09
Specifications
| Product Type | Flame-resistant Coveralls |
|---|---|
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Disposable | Reusable |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-200-857
|
DuPont™
ST127SWH2X002500 |
2X-Large |
Case of 25 Each for $309.31
|
|
|||||
|
17-200-858
|
DuPont™
ST127SWH3X002500 |
3X-Large |
Case of 25 Each for $327.86
|
|
|||||
|
17-200-859
|
DuPont™
ST127SWH4X002500 |
4X-Large |
Case of 25 Each for $354.07
|
|
|||||
|
17-200-860
|
DuPont™
ST127SWH5X002500 |
5X-Large |
Case of 25 Each for $382.43
|
|
|||||
|
17-200-861
|
DuPont™
ST127SWH6X002500 |
6X-Large |
Case of 25 Each for $412.99
|
|
|||||
|
17-200-862
|
DuPont™
ST127SWH7X002500 |
7X-Large |
Case of 25 Each for $446.09
|
|
|||||
|
17-111-309
|
DuPont™
ST127SWHLG002500 |
Large |
Case of 25 Each for $291.85
|
|
|||||
|
17-111-308
|
DuPont™
ST127SWHMD002500 |
Medium |
Case of 25 Each for $291.85
|
|
|||||
|
17-111-310
|
DuPont™
ST127SWHXL002500 |
X-Large |
Case of 25 Each for $291.85
|
|
|||||
Description
- Garments are lightweight, disposable secondary flame-resistant overgarments designed to help protect and preserve primary flame-resistant garments, while providing an ideal balance of protection, durability, and comfort.
- In the event that a flash fire occurs, Tyvek™ 400 SFR garments won't ignite and won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
- Composed of flash spun high density polyethylene breathable nonwoven Tyvek™ fabric that has been flame-retardant treated, available only from DuPont, Tyvek™ 400 SFR garments offer an inherent barrier against particles (down to 1.0 micron in size).
- Tyvek™ 400 SFR represents a new generation of disposable secondary flame-resistant garment technology.
- Can be recycled through the Tyvek™ protective apparel recycling program if not contaminated with hazardous materials.
- Applications include: oil and gas, general manufacturing/operations, maintenance, dirty jobs, environmental services, utilities.
- Relaxed fit coverall design features a diamond gusset crotch to allow for unrestricted movement and optimal fit when wearing primary flame-resistant protective clothing beneath
- Red trim elements - red external stitched seams, red zipper, and red chest patch border
- Patent-pending design
- Attached standard-fit hood with elastic around face opening
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Storm flap
Specifications
| Flame-resistant Coveralls | |
| Tyvek | |
| Reusable | |
| Elastic | |
| Hooded | |
| Zipper | |
| Non-sterile | |
| None |
| Unisex | |
| White | |
| Standard | |
| Elastic | |
| Attached | |
| Serged | |
| Flame |