missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 SFR White Coverall ST120, Standard Fit Hood

Catalog No. 17200859
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
Large
Medium
X-Large
Catalog No. Size
17200859 4X-Large
17200857 2X-Large
17200858 3X-Large
17200860 5X-Large
17200861 6X-Large
17200862 7X-Large
17111309 Large
17111308 Medium
17111310 X-Large
9 options

Catalog No. 17200859

Supplier: DuPont™ ST127SWH4X002500

Only null left

Tyvek™ 400 SFR Coverall. Relaxed Fit Design. Standard Fit Hood. Storm flap. Red Zipper. Elastic Wrists and Ankles. Red External Serged Seams. White.

  • Garments are lightweight, disposable secondary flame-resistant overgarments designed to help protect and preserve primary flame-resistant garments, while providing an ideal balance of protection, durability, and comfort.
  • In the event that a flash fire occurs, Tyvek™ 400 SFR garments won't ignite and won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
  • Composed of flash spun high density polyethylene breathable nonwoven Tyvek™ fabric that has been flame-retardant treated, available only from DuPont, Tyvek™ 400 SFR garments offer an inherent barrier against particles (down to 1.0 micron in size).
  • Tyvek™ 400 SFR represents a new generation of disposable secondary flame-resistant garment technology.
  • Can be recycled through the Tyvek™ protective apparel recycling program if not contaminated with hazardous materials.
  • Applications include: oil and gas, general manufacturing/operations, maintenance, dirty jobs, environmental services, utilities.
  • Relaxed fit coverall design features a diamond gusset crotch to allow for unrestricted movement and optimal fit when wearing primary flame-resistant protective clothing beneath
  • Red trim elements - red external stitched seams, red zipper, and red chest patch border
  • Patent-pending design
  • Attached standard-fit hood with elastic around face opening
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Storm flap

Specifications

Product Type Flame-resistant Coveralls
Size 4X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Reusable
Waist Style Standard
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Flame
No. of Pockets None
Show More Show Less