missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 400 SFR White Coverall ST120, Collared
Tyvek™ 400 SFR Coverall. Relaxed Fit Design. Collar. Storm flap. Red Zipper. Elastic Wrists and Ankles. Red External Serged Seams. White.
Supplier: DuPont™ ST120SWH4X002500
Description
- Garments are lightweight, disposable secondary flame-resistant overgarments designed to help protect and preserve primary flame-resistant garments, while providing an ideal balance of protection, durability, and comfort.
- In the event that a flash fire occurs, Tyvek™ 400 SFR garments won't ignite and won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
- Composed of flash spun high density polyethylene breathable nonwoven Tyvek™ fabric that has been flame-retardant treated, available only from DuPont, Tyvek™ 400 SFR garments offer an inherent barrier against particles (down to 1.0 micron in size).
- Tyvek™ 400 SFR represents a new generation of disposable secondary flame-resistant garment technology. Compared to traditional disposable secondary flame-resistant garments that have been available for years, Tyvek™ 400 SFR is a differentiated multi-hazard garment solution, offering superior particle barrier protection and improved durability in a lighter-weight fabric.
- Can be recycled through the Tyvek™ protective apparel recycling program if not contaminated with hazardous materials.
- Applications include: oil and gas, general manufacturing/operations, maintenance, dirty jobs, environmental services, utilities.
Specifications
| Flame-resistant Coveralls | |
| Unisex | |
| White | |
| Standard | |
| Elastic | |
| Standard Collar | |
| Serged | |
| Flame |
| 4X-Large | |
| Tyvek | |
| Reusable | |
| Elastic | |
| Collared | |
| Zipper | |
| Non-sterile | |
| None |