missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Hooded Coverall
GREENER_CHOICE

Catalog No. 19820534
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820534 Large
19820535 Medium
19820536 X-Large
19820528 2X-Large
19820529 3X-Large
19820530 4X-Large
19820531 5X-Large
19820532 6X-Large
19820533 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19820534 Supplier DuPont™ Supplier No. TY127SWHLG0025VP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash. Individually packaged for PPE vending machines.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps.

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Non-toxic Particulates
Certifications/Compliance ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
Latex Free Yes
Model TY127SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Height (English) 5.5 to 5.9 ft.
Quantity 25/Cs.
Show More Show Less