missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 400 Hooded Coverall
Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash. Individually packaged for PPE vending machines.
Supplier: DuPont™ TY127SWH6X0025VP
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment
- Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
- Longer zipper extends to chin for complete coverage of neck area
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
- Stormflaps.
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Elastic | |
| Respirator Fit | |
| Serged | |
| ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127 | |
| TY127SWH | |
| 6.9 to 7.1 ft. |
| 6X-Large | |
| Tyvek | |
| Limited-use | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| Yes | |
| Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning | |
| 25/Cs. |