missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots, Vending Packaged
GREENER_CHOICE

Catalog No. 19820517
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820517 Medium
19820516 Large
19820518 X-Large
19820510 2X-Large
19820511 3X-Large
19820512 4X-Large
19820513 5X-Large
19820514 6X-Large
19820515 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19820517 Supplier DuPont™ Supplier No. TY122SWHMD0025VP

Coveralls with attached skid-resistant boots include stormflaps and a respirator fit hood. Individually packaged for PPE vending machines.

  • Comfort fit design based on extensive wearer input, individually packaged for PPE vending machine use
  • Enables a greater range of movement while stretching and bending
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Attached Tyvek™ boots have special coating to provide added skid-resistance
  • 25 Coveralls/Cs.

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic, Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Model TY122SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance and Operations, Spray Painting, General Cleaning
Packaging Individually Packaged for PPE Vending
Show More Show Less