Learn More
DuPont™ Tyvek™ 400 D Dual Coverall
Suitable for applications where comfort requirements are combined with limited protective requirements for frontal exposures.
Supplier:  DuPont™ TD125SWBLG0025CM
 View more versions of this product
Catalog No. 19-716-102
Only null left
Explore available promotions
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex
  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar
  • Longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrist
  • Thumb loop to keep garment sleeve in place
  • Elastic opening for tighter fit at ankle
Compliance
Meets USDA, FDA, OSHA standards; ASTM D751, ASTM D1117, ASTM D5034, ASTM D257; AATCC 127 compliant
Specifications
Specifications
General Purpose Coveralls
Unisex
White, Blue
Elastic
Collared
Front Zipper
Non-sterile
USDA, FDA, OSHA, ASTM D751, ASTM D1117, ASTM D5034, ASTM D257, AATCC 127
39.75 to 43.25 in.
50 in.
127 cm
5.7 mil
35 in.
26.5 in.
5.5 to 5.9 ft.
Large
Tyvek, SMS
Single-use
Elastic
Unhooded
Serged
Laydown
TD125SWB
29 in.
101 to 110 cm
Full Length with Thumb Loop
Fiberglass and Composite Manufacturing, Wind Turbines, Boat Construction and Repair, Remediation, Maintenance
89 cm
67.35 cm
167.64 to 179.83 cm
Product Suggestions
Product Suggestions